Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007984.2 | NP_032010.2 | 2 | 957 | ||
GenBank mRNA | AK083935.1 | - | 2 | 958 | ||
AK167670.1 | - | 2 | 957 | |||
BC010338.1 | - | 2 | 943 | |||
BC029281.1 | - | 2 | 945 | |||
BC037137.1 | - | 2 | 954 | |||
BC052408.1 | - | 2 | 946 | 5716179 | ||
L33726.1 | - | 2 | 941 |