Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008024.2 | NP_032050.2 | 1 | 1027 | ||
GenBank mRNA | AB221616.1 | - | 1 | 750 | ||
AK050949.1 | - | 1 | 249 | |||
BC137805.1 | - | 1 | 772 | 8860824 | ||
BC137806.1 | - | 1 | 772 | 8860825 |