Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159518.1 | NP_001152990.1 | 2, 3 | 607 | ||
NM_008048.3 | NP_032074.3 | 1, 2 | 514 | |||
GenBank mRNA | AB012886.1 | - | 1, 2 | 515 | ||
AK002277.1 | - | 1, 2 | 510 | |||
AK148749.1 | - | 1, 2 | 511 | |||
AK156315.1 | - | 1, 2 | 516 | |||
BC047202.1 | - | 1, 2 | 426 | |||
BC092538.1 | - | 1, 2 | 510 | 30284911 | ||
BF471478.1 | - | 2, 3 | 121 | |||
L75822.2 | - | 1, 2 | 493 |