Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160406.1 | NP_001153878.1 | 4 | 638 | ||
NM_008114.3 | NP_032140.1 | 4 | 638 | |||
GenBank mRNA | AF017275.1 | - | 4 | 597 | ||
AK157262.1 | - | 4 | 647 | |||
BC052654.1 | - | 4 | 613 | 30039774 | ||
BC132365.1 | - | 4 | 574 | 40130642 | ||
BC132367.1 | - | 4 | 574 | 40130644 | ||
BC145238.1 | - | 4 | 574 | 9053790 |