Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159518.1 | NP_001152990.1 | 3 | 660 | ||
NM_008048.3 | NP_032074.3 | 2 | 567 | |||
GenBank mRNA | AB012886.1 | - | 2 | 568 | ||
AK002277.1 | - | 2 | 563 | |||
AK148749.1 | - | 2 | 564 | |||
AK156315.1 | - | 2 | 569 | |||
AW318969.1 | - | 2 | 562 | |||
BC047202.1 | - | 2 | 479 | |||
BC092538.1 | - | 2 | 563 | 30284911 | ||
BF471478.1 | - | 3 | 174 | |||
L75822.2 | - | 2 | 546 |