Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008077.4 | NP_032103.2 | 5 | 686 | ||
GenBank mRNA | AF326547.1 | - | 5 | 636 | ||
AF326548.1 | - | 5 | 636 | |||
AF483492.1 | - | 4 | 453 | |||
AF483493.1 | - | 4 | 453 | |||
AK047521.1 | - | 5 | 822 | |||
AK054554.1 | - | 5 | 688 | |||
AK136801.1 | - | 5 | 671 | |||
AK148497.1 | - | 5 | 686 | |||
AK158827.1 | - | 4 | 452 | |||
AK160868.1 | - | 5 | 684 | |||
BC027059.1 | - | 5 | 637 | 5358787 | ||
KC134210.1 | - | 4 | 453 | |||
KM102984.1 | - | 5 | 686 | |||
KM102985.1 | - | 5 | 821 | |||
Y12257.1 | - | 4 | 453 | |||
Z49976.1 | - | 5 | 573 |
Gene Symbol: Gad1-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137453.1 | - | 1 | 762 | ||
AK137640.1 | - | 1 | 786 |