Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008089.2 | NP_032115.1 | 6 | 1415 | ||
GenBank mRNA | AK146915.1 | - | 6 | 1400 | ||
BC052653.1 | - | 6 | 1382 | 30039400 | ||
X15763.1 | - | 6 | 1320 |