Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008098.4 | NP_032124.1 | 4 | 766 | ||
GenBank mRNA | AK002620.1 | - | 4 | 680 | ||
AK028027.1 | - | 4 | 694 | |||
AK049398.1 | - | 4 | 666 | |||
AK083888.1 | - | 4 | 680 | |||
AK132029.1 | - | 4 | 689 | |||
AK145755.1 | - | 4 | 671 | |||
AK146305.1 | - | 4 | 714 | |||
AK146473.1 | - | 4 | 666 | |||
AK150750.1 | - | 4 | 688 | |||
AK151106.1 | - | 4 | 634 | |||
AK151147.1 | - | 4 | 634 | |||
AK151485.1 | - | 4 | 691 | |||
AK151564.1 | - | 4 | 634 | |||
AK151640.1 | - | 4 | 689 | |||
AK151687.1 | - | 4 | 662 | |||
AK152272.1 | - | 4 | 662 | |||
AK152317.1 | - | 4 | 651 | |||
AK191828.1 | - | 1 | 21 | |||
BC043084.1 | - | 4 | 663 | 6417343 | ||
BC054811.1 | - | 4 | 679 | 6839739 | ||
D78188.1 | - | 4 | 675 | |||
U20290.1 | - | 4 | 697 |