Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160406.1 | NP_001153878.1 | 7 | 1747 | ||
NM_008114.3 | NP_032140.1 | 7 | 1648 | |||
GenBank mRNA | AF017275.1 | - | 7 | 1606 | ||
AK157262.1 | - | 7 | 1657 | |||
BB734623.1 | - | 1 | 85 | |||
BC052654.1 | - | 7 | 1623 | 30039774 |