Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008130.2 | NP_032156.2 | 2, 3 | 565 | ||
GenBank mRNA | AK144851.1 | - | 2, 3 | 349 | ||
AK158261.1 | - | 2, 3 | 565 | |||
BC141135.1 | - | 2, 3 | 266 | 9055747 | ||
BC145445.1 | - | 2, 3 | 266 | 9054003 | ||
X95255.1 | - | 2, 3 | 305 |