Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008221.4 | NP_032247.1 | 1, 2 | 155 | ||
GenBank mRNA | AK011619.1 | - | 1, 2 | 156 | ||
AK012501.1 | - | 1, 2 | 157 | |||
AK012897.1 | - | 1, 2 | 156 | |||
BC057014.1 | - | 1, 2 | 132 | 6817810 | ||
M26897.1 | - | 1, 2 | 117 | |||
V00857.1 | - | 1, 2 | 37 |