Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008137.4 | NP_032163.3 | 2 | 1242 | ||
GenBank mRNA | AK035320.1 | - | 1 | 82 | ||
AK085444.1 | - | 2 | 1148 | |||
AK160808.1 | - | 2 | 836 | |||
AK168498.1 | - | 2 | 898 | |||
BC027015.1 | - | 2 | 710 | 5375817 | ||
M80631.1 | - | 2 | 371 |