Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163465.1 | NP_001156937.1 | 3, 4 | 438 | ||
NM_008164.2 | NP_032190.2 | 3, 4 | 438 | |||
GenBank mRNA | AK143132.1 | - | 3, 4 | 419 | ||
AK220431.1 | - | 3, 4 | 355 | |||
BC004843.1 | - | 3, 4 | 408 | |||
BC052010.1 | - | 3, 4 | 397 | 5717786 | ||
U43194.1 | - | 3, 4 | 332 |