Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008182.3 | NP_032208.2 | 7 | 750 | ||
GenBank mRNA | AK002712.1 | - | 7 | 757 | ||
BC061133.1 | - | 6 | 693 | 30303519 | ||
J03958.1 | - | 7 | 765 |