Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008130.2 | NP_032156.2 | 14 | 2784 | ||
GenBank mRNA | BC141135.1 | - | 14 | 2485 | 9055747 | |
BC145445.1 | - | 14 | 2485 | 9054003 | ||
CA752350.1 | - | 3 | 642 | |||
X95255.1 | - | 14 | 2524 |