Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160016.1 | NP_001153488.1 | 12 | 1470 | ||
NM_001160017.1 | NP_001153489.1 | 11 | 1421 | |||
NM_008142.4 | NP_032168.1 | 12 | 1473 | |||
GenBank mRNA | AB246709.1 | - | 10 | 1167 | ||
AB246710.1 | - | 10 | 1164 | |||
AK156101.1 | - | 11 | 1418 | |||
AK163844.1 | - | 12 | 1462 | |||
U29055.1 | - | 10 | 1200 |