Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008236.4 | NP_032262.2 | 5 | 500 | ||
GenBank mRNA | AK053792.1 | - | 5 | 501 | ||
BC138112.1 | - | 5 | 828 | 8861131 |