Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Onecut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008262.3 | NP_032288.1 | 1 | 1243 | ||
GenBank mRNA | BC024053.1 | - | 1 | 1243 | 5101697 | |
U95945.1 | - | 1 | 1036 |