Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008268.2 | NP_032294.2 | 2 | 1322 | ||
GenBank mRNA | AK133512.1 | - | 2 | 1238 | ||
BC040755.1 | - | 2953 | ||||
BC103595.1 | - | 2 | 1242 | 40046040 | ||
BC103596.1 | - | 2 | 1242 | 40046042 | ||
BC103604.1 | - | 2 | 1242 | 40046047 | ||
BC103607.1 | - | 2 | 1242 | 40046044 | ||
M26283.1 | - | 2 | 1322 |