Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170853.1 | NP_001164324.1 | 414 | |||
GenBank mRNA | AK087931.1 | - | 2 | 243 | ||
AK136391.1 | - | 415 | ||||
FJ594740.1 | - | 2 | 313 | |||
FJ594741.1 | - | 322 |
Gene Symbol: Ifi203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022371.1 | - | 475 | |||
AK040214.1 | - | 4 | 415 | |||
AK041232.1 | - | 4 | 403 | |||
AK137616.1 | - | 4 | 414 | |||
AK144663.1 | - | 4 | 396 | |||
AK161531.1 | - | 2 | 258 | |||
AK164695.1 | - | 2 | 252 | |||
AK172505.1 | - | 489 | ||||
BC008167.1 | - | 517 | 3257492 | |||
BE632791.1 | - | 2 | 193 | |||
CA574467.1 | - | 2 | 131 |