Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008252.3 | NP_032278.1 | 5 | 863 | ||
GenBank mRNA | AK003773.1 | - | 5 | 769 | ||
AK008443.1 | - | 5 | 774 | |||
AK012568.1 | - | 5 | 830 | |||
AK014053.1 | - | 1 | 66 | |||
AK076075.1 | - | 5 | 799 | |||
AK135296.1 | - | 5 | 769 | |||
AK135297.1 | - | 5 | 769 | |||
AK146212.1 | - | 5 | 798 | |||
AK153858.1 | - | 5 | 772 | |||
BC002050.1 | - | 5 | 743 | 3489780 | ||
BC046759.1 | - | 5 | 767 | 5693598 | ||
BC083108.1 | - | 5 | 761 | 5320702 | ||
Z46757.1 | - | 5 | 772 |