Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008306.4 | NP_032332.2 | 9 | 2085 | ||
GenBank mRNA | AF049894.1 | - | 9 | 1931 | ||
AF074926.1 | - | 9 | 1931 | |||
AK147282.1 | - | 10 | 2478 | |||
BC066098.1 | - | 9 | 2085 | 30533342 | ||
BC079561.1 | - | 9 | 2085 | 30533342 |