Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 2 | 355 | ||
GenBank mRNA | AK040399.1 | - | 2 | 139 | ||
AK041307.1 | - | 2 | 356 | |||
AK041403.1 | - | 57 | ||||
AK042009.1 | - | 3 | 1024 | |||
AK052452.1 | - | 3 | 1043 | |||
AK139858.1 | - | 3 | 716 | |||
AK145184.1 | - | 2 | 646 | |||
BC110553.2 | - | 2 | 264 | 40047584 | ||
X07962.1 | - | 2 | 655 |