Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008380.1 | NP_032406.1 | 2 | 315 | ||
GenBank mRNA | AK134991.1 | - | 4 | 766 | ||
AK135474.1 | - | 1 | 147 | |||
BC053527.1 | - | 2 | 308 | 6528822 | ||
CT010380.1 | - | 1 | 143 | |||
X69619.1 | - | 2 | 315 |