Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008408.4 | NP_032434.3 | 4, 5 | 332 | ||
GenBank mRNA | AK030363.1 | - | 4, 5 | 396 | ||
AK077877.1 | - | 4, 5 | 356 | |||
AK081547.1 | - | 4, 5 | 388 | |||
AK087470.1 | - | 4, 5 | 371 | |||
AK129027.1 | - | 4, 5 | 370 | |||
AK150640.1 | - | 4, 5 | 377 | |||
AK151541.1 | - | 4, 5 | 388 | |||
AK151791.1 | - | 4, 5 | 388 | |||
AK152242.1 | - | 4, 5 | 377 | |||
AK153840.1 | - | 4, 5 | 333 | |||
AK153886.1 | - | 4, 5 | 433 | |||
AK160037.1 | - | 4, 5 | 371 | |||
AK160607.1 | - | 4, 5 | 333 | |||
AK168505.1 | - | 4, 5 | 377 | |||
AK168937.1 | - | 4, 5 | 377 | |||
AK169669.1 | - | 4, 5 | 356 | |||
AX799090.1 | - | 4, 5 | 375 | |||
BC028897.1 | - | 3, 4 | 283 | |||
BC037612.1 | - | 4, 5 | 371 | 5353797 | ||
BC085313.1 | - | 4, 5 | 361 | 30443880 | ||
L34260.1 | - | 4, 5 | 375 |