Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008444.4 | NP_032470.3 | 2 | 233 | ||
GenBank mRNA | AK081541.1 | - | 2 | 190 | ||
AK083700.1 | - | 2 | 221 | |||
AK122258.1 | - | 2 | 194 | |||
AK147467.1 | - | 2 | 227 | |||
AK148200.1 | - | 2 | 214 | |||
BC036410.1 | - | 1 | 146 | |||
BC065132.1 | - | 2 | 200 | |||
BC139388.1 | - | 1 | 152 | 8862410 | ||
BC139389.1 | - | 1 | 152 | 8862411 | ||
CJ089315.1 | - | 2 | 236 | |||
D26077.1 | - | 1 | 146 |