Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008403.4 | NP_032429.1 | 4 | 449 | ||
GenBank mRNA | AJ001373.1 | - | 4 | 431 | ||
AK002786.1 | - | 4 | 406 | |||
AK004172.1 | - | 4 | 405 | |||
AK007635.1 | - | 4 | 407 | |||
AK075577.1 | - | 4 | 450 | |||
AK089937.1 | - | 4 | 563 | |||
AK147929.1 | - | 4 | 398 | |||
BC021151.1 | - | 4 | 588 | |||
BC028772.1 | - | 3 | 285 | 1230640 |