Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008418.2 | NP_032444.2 | 1 | 1237 | ||
GenBank mRNA | BC132349.1 | - | 1 | 1074 | 40130626 | |
BC137667.1 | - | 1 | 1074 | 8860686 |