Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008435.2 | NP_032461.2 | 4 | 1936 | ||
GenBank mRNA | AF008573.1 | - | 4 | 1913 | ||
AK140377.1 | - | 1 | 1180 | |||
AK158889.1 | - | 2 | 1489 |