Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271704.1 | NP_001258633.1 | 2 | 1515 | ||
NM_181317.4 | NP_851834.1 | 2 | 1883 | |||
GenBank mRNA | AK048819.1 | - | 2 | 1857 | ||
AK053281.1 | - | 1 | 288 | |||
AK087481.1 | - | 2 | 1563 | |||
AK173110.1 | - | 2 | 1883 | |||
BC059833.1 | - | 2 | 1515 | 6838254 |