Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 3 | 366 | ||
GenBank mRNA | AK028259.1 | - | 3 | 367 | ||
AK028307.1 | - | 3 | 385 | |||
AK154800.1 | - | 3 | 384 | |||
AK163411.1 | - | 191 | ||||
AK169311.1 | - | 3 | 389 | |||
AK169348.1 | - | 3 | 389 | |||
AK169464.1 | - | 3 | 405 | |||
AK195236.1 | - | 2 | 110 | |||
AK218219.1 | - | 2 | 126 | |||
BC006771.1 | - | 3 | 351 | 3588534 | ||
U20619.1 | - | 3 | 371 | |||
U34228.1 | - | 2 | 215 |