Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008483.3 | NP_032509.2 | 3, 4 | 380 | ||
GenBank mRNA | AK140351.1 | - | 3, 4 | 359 | ||
AK158444.1 | - | 3, 4 | 352 | |||
BC026051.1 | - | 3, 4 | 307 | 4036357 |