Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lta4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008517.2 | NP_032543.2 | 2 | 363 | ||
GenBank mRNA | AK032828.1 | - | 2 | 308 | ||
AK134931.1 | - | 2 | 349 | |||
BC021417.1 | - | 2 | 357 | 5004113 | ||
M63848.1 | - | 329 |