Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008476.3 | NP_032502.3 | 9 | 1566 | ||
GenBank mRNA | AK009394.1 | - | 9 | 1569 | ||
AK010039.1 | - | 4 | 459 | |||
BC064470.1 | - | 8 | 1680 | |||
BC080820.1 | - | 9 | 1566 | 30141946 | ||
K02108.1 | - | 9 | 1558 |