Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005109.2 | NP_005100.1 | 2 | 470 | ||
XM_005265638.2 | XP_005265695.1 | 2 | 474 | |||
XM_011534331.2 | XP_011532633.1 | 2 | 522 | |||
XM_017007601.1 | XP_016863090.1 | 2 | 313 | |||
XM_017007602.1 | XP_016863091.1 | 2 | 474 | |||
GenBank mRNA | AB017642.1 | - | 2 | 470 | ||
AB029024.1 | - | 2 | 227 | |||
AK309634.1 | - | 2 | 358 | |||
BC008726.2 | - | 2 | 453 | 3163379 |