Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008536.3 | NP_032562.1 | 5, 6 | 1017 | ||
GenBank mRNA | AK077326.1 | - | 3, 4 | 520 | ||
AK133556.1 | - | 518 | ||||
AK134603.1 | - | 4, 5 | 953 | |||
AK164718.1 | - | 3, 4 | 521 | |||
BC083073.1 | - | 5, 6 | 1017 | 6337376 | ||
KC508089.1 | - | 3, 4 | 492 | |||
L15429.1 | - | 3, 4 | 507 | |||
L15443.1 | - | 3, 4 | 507 |