Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008569.2 | NP_032595.2 | 2 | 461 | ||
GenBank mRNA | AK052404.1 | - | 2 | 442 | ||
AK053383.1 | - | 1 | 1534 | |||
AK054305.1 | - | 2 | 443 | |||
AK077847.1 | - | 2 | 462 | |||
AK090134.1 | - | 2 | 444 | |||
AK149762.1 | - | 2 | 433 | |||
AK164804.1 | - | 2 | 447 | |||
BC057201.1 | - | 2 | 420 | |||
BC139002.1 | - | 2 | 442 | 8862024 | ||
BC145492.1 | - | 2 | 442 | 9054055 | ||
X80169.1 | - | 2 | 446 |