Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008587.1 | NP_032613.1 | 3 | 576 | ||
GenBank mRNA | U21301.1 | - | 3 | 576 |