Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008653.2 | NP_032679.2 | 2 | 307 | ||
GenBank mRNA | AF097333.1 | - | 2 | 258 | ||
AK146661.1 | - | 2 | 308 | |||
AK146886.1 | - | 2 | 308 | |||
AK169302.1 | - | 2 | 288 | |||
AK169306.1 | - | 2 | 288 | |||
BC131672.1 | - | 2 | 283 | |||
BC154408.1 | - | 2 | 328 | 40110890 |