Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033341.4 | NP_203127.3 | 1 | 1412 | ||
GenBank mRNA | AF420440.1 | - | 1 | 1411 | ||
BC039318.1 | - | 1 | 1182 | 5269405 | ||
BC046168.1 | - | 1 | 1156 | |||
BC071665.1 | - | 1 | 960 | 5171408 |