Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256005.1 | NP_001242934.1 | 11 | 1838 | ||
NM_008620.3 | NP_032646.2 | 11 | 1775 | |||
GenBank mRNA | AK220567.1 | - | 11 | 1756 | ||
BC141364.1 | - | 11 | 1726 | 9055977 | ||
BC147434.1 | - | 11 | 1726 | 9055976 | ||
EF494423.1 | - | 11 | 1838 | |||
EF494424.1 | - | 11 | 1809 |