Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093775.1 | NP_001087244.1 | 17, 18 | 3548 | ||
NM_001093776.1 | NP_001087245.1 | 17, 18 | 3454 | |||
NM_001093778.1 | NP_001087247.1 | 17, 18 | 3539 | |||
NM_008666.3 | NP_032692.2 | 17, 18 | 3542 | |||
GenBank mRNA | AB093283.1 | - | 14, 15 | 2901 | ||
AB212898.1 | - | 16, 17 | 3146 | |||
AF004295.1 | - | 17, 18 | 3358 | |||
AK049967.1 | - | 8, 9 | 1383 | |||
AK076608.1 | - | 5, 6 | 979 | |||
AK161925.1 | - | 1, 2 | 83 | |||
BC079651.1 | - | 17, 18 | 3423 | |||
BC094438.1 | - | 13, 14 | 2779 | 30692780 | ||
BC131677.1 | - | 16, 17 | 3143 | 40111036 | ||
U86338.1 | - | 17, 18 | 3270 |