Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008707.3 | NP_032733.1 | 1 | 82 | ||
GenBank mRNA | AF043326.1 | - | 1 | 94 | ||
AK146523.1 | - | 1 | 97 | |||
BC016526.1 | - | 1 | 82 | 3589678 | ||
BC021635.1 | - | 1 | 96 | 3989540 |