Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008760.4 | NP_032786.1 | 2 | 240 | ||
GenBank mRNA | AK028300.1 | - | 2 | 266 | ||
AK053650.1 | - | 2 | 241 | |||
AK075638.1 | - | 1 | 197 | |||
AK160760.1 | - | 2 | 262 | |||
AK163772.1 | - | 1 | 193 | |||
BC021939.1 | - | 1 | 143 | 5067073 | ||
D31951.1 | - | 1 | 153 |