Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008764.3 | NP_032790.3 | 2 | 415 | ||
GenBank mRNA | AB013898.1 | - | 2 | 415 | ||
AK140244.1 | - | 2 | 238 | |||
AK146105.1 | - | 2 | 358 | |||
BC049782.1 | - | 2 | 232 | 6506550 | ||
U94331.1 | - | 2 | 268 |