Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008730.2 | NP_032756.1 | 3 | 965 | ||
GenBank mRNA | AK140128.1 | - | 3 | 482 | ||
BC093479.1 | - | 3 | 890 | 30947054 | ||
BC139018.1 | - | 3 | 872 | 8862040 | ||
BC139020.1 | - | 3 | 872 | 8862042 |