Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008695.2 | NP_032721.2 | 21 | 4319 | ||
GenBank mRNA | AB017202.1 | - | 21 | 4345 | ||
AK004203.1 | - | 2 | 65 | |||
AK051664.1 | - | 21 | 4299 | |||
AK083629.1 | - | 21 | 4296 | |||
AK128978.1 | - | 19 | 3540 | |||
AK140832.1 | - | 21 | 4293 | |||
AK164258.1 | - | 21 | 4290 | |||
AY074820.1 | - | 21 | 4280 | |||
BC054746.1 | - | 21 | 4301 | 6405073 | ||
BC057016.1 | - | 21 | 4247 | 5719452 |