Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190187.1 | NP_001177116.1 | 10 | 2478 | ||
NM_001190188.1 | NP_001177117.1 | 9 | 2433 | |||
NM_008734.3 | NP_032760.1 | 10 | 2481 | |||
GenBank mRNA | AF010130.1 | - | 10 | 2481 | ||
AK014399.1 | - | 3 | 702 |