Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008740.4 | NP_032766.2 | 14 | 1574 | ||
GenBank mRNA | AK028415.1 | - | 3 | 298 | ||
AK049281.1 | - | 14 | 1544 | |||
AK085086.1 | - | 14 | 1528 | |||
AK153905.1 | - | 14 | 1543 | |||
AK208793.1 | - | 3 | 276 | |||
AK213251.1 | - | 3 | 297 | |||
AK214505.1 | - | 3 | 216 | |||
BC006627.1 | - | 14 | 1579 | 3968733 | ||
BC019167.1 | - | 14 | 1527 | 4506351 | ||
U10120.1 | - | 14 | 1532 |