Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008810.2 | NP_032836.1 | 3 | 209 | ||
GenBank mRNA | AK145718.1 | - | 3 | 287 | ||
AK148459.1 | - | 3 | 230 | |||
AK167987.1 | - | 3 | 235 | |||
AK168152.1 | - | 3 | 259 | |||
AK169289.1 | - | 3 | 284 | |||
AK212590.1 | - | 2 | 130 | |||
BC007142.1 | - | 3 | 209 | 3600605 | ||
M76727.1 | - | 3 | 296 |